8-Quinolineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Quinolin-8-ylboronic acid

BB-5183-1G BB-5183-5G
COBBBB-5183-1GEA 0 GBP
COBBBB-5183-1G COBBBB-5183-5G
8-Quinolineboronic acid
8-Quinolineboronic acid
Formula: C₉H₈BNO₂
MW: 172.98 g/mol
MDL Number: MFCD01114698
CAS Number: 86-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR