6-Quinolineboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Quinolin-6-ylboronic acid, Quinoline-6-boronic acid
BB-5182-1G
BB-5182-5G
COBBBB-5182-1GEA
0
GBP
COBBBB-5182-1G
COBBBB-5182-5G
6-Quinolineboronic acid
6-Quinolineboronic acid
Formula:
C₉H₈BNO₂ MW: 172.98 g/mol |
MDL Number:
MFCD03093100 CAS Number: 376581-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...