4-Quinolineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Quinolin-4-ylboronic acid

BB-8392-5G BB-8392-1G
COBBBB-8392-5GEA 0 GBP
COBBBB-8392-5G COBBBB-8392-1G
4-Quinolineboronic acid
4-Quinolineboronic acid
Formula: C₉H₈BNO₂
MW: 172.9763 g/mol
MDL Number: MFCD03095174
CAS Number: 371764-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR