SIALD5905

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SIALD5905-50TABEA 543 GBP
SIALD5905-50TAB SIALD5905-100TAB
SIALD5905
10 mg substrate per tablet

Application: Reagent for spectrophotometric determination of selenium.

Reconstitution: Dissolve 1 tablet in 15 ml of Tris-buffered saline, pH 7.6. Add 12 μl of fresh 30% hydrogen peroxide. DAB and hydrogen peroxide concentration may be adjusted to suit individual applications.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR