N-Decanoyl-N-methyl-D-glucamine (MEGA-10)

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

MOLE22118459-1GEA 0 GBP
MOLE22118459-1G
N-Decanoyl-N-methyl-D-glucamine (MEGA-10)
N-Decanoyl-N-methyl-D-glucamine (MEGA-10)
Formula: C₁₇H₃₅NO₆
MW: 349.46 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036801
CAS Number: 85261-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR