(4-Nitrophenyl)-β-D-galactopyranoside

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: pNPG, 4-Nitrophenyl β-D-Galactopyranoside

4013562.0005 4013562.0025
BACE4013562.0005EA 0 GBP
BACE4013562.0005 BACE4013562.0025
(4-Nitrophenyl)-β-D-galactopyranoside
(4-Nitrophenyl) β-D-galactopyranoside
Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063256
CAS Number: 3150-24-1
EINECS: 221-584-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR