Brilliant green ≥90% (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®

Supplier: SIGMA-ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: CI 42040Basic Green 1Brilliant Green hydrogen sulphateDiamond GreenAstradiamant green GXN-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphateSolid Green JOEthyl GreenMalachite Green GEmerald Green

M-Clarity™ quality level = MQ200

B4014-25GEA 145 GBP
B4014-25G
Brilliant green ≥90% (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
Brilliant Green

Certified for bacteriostatic potency. Tested as a bacteriostatic agent against a spore-former and two members of the colon-typhoid group.

Formula: C₂₇H₃₄N₂O₄S
MW: 482.65 g/mol
Melting Pt: 210 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011880
CAS Number: 633-03-4
EINECS: 211-190-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR