1,2-Bis(dichlorophosphino)ethane 96%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
B21092.03
B21092.06
B21092.14
B21092.03EA
60.4
GBP
B21092.03
B21092.06
B21092.14
1,2-Bis(dichlorophosphino)ethane 96%
1,2-Bis(dichlorophosphino)ethane
Purity: 96%
CAS No.: 28240-69-9
Molecular Formula: Cl2PCH2CH2PCl2
Formula Weight: 231.81
Flash Point: None
Refractive Index: 1.5880
Boiling Point: 70°/1mm
UN No.: 3265
MDL No.: MFCD00009609
Formula:
C₂H₄Cl₄P₂ Boiling Pt: 70 °C (1 mmHg) Density: 1.54 Storage Temperature: Ambient |
MDL Number:
MFCD00009609 CAS Number: 28240-69-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...