2-Chloro-4-methylanisole

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-1-methoxy-4-methylbenzene, 3-Chloro-4-methoxytoluene

C2291.10 C2291.50
XXXXC2291.10EA 0 GBP
XXXXC2291.10 XXXXC2291.50
2-Chloro-4-methylanisole
2-Chloro-4-methylanisole
Formula: C₈H₉ClO
MW: 156.609 g/mol
MDL Number: MFCD00060649
CAS Number: 22002-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR