N-Acetyl-β-D-glucosamine

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine

ART-101 ART-101-1 ART-101-2
BTRNART-101EA 0 GBP
BTRNART-101 BTRNART-101-1 BTRNART-101-2
N-Acetyl-β-D-glucosamine
N-Acetyl-β-D-glucosamine
Formula: C₈H₁₅NO₆
MW: 221.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00136044
CAS Number: 7512-17-6
EINECS: 231-368-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR