β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
((2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy-((((2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy)hydroxyphosphoryl)oxyphosphinic acid disodium salt
, Nicotinamide adenine dinucleotide reduced, disodium salt (reduced form)
, NADH disodium salt
SIALN7410-15VLEA
296
GBP
SIALN7410-15VL
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), Sigma-Aldrich®
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)
Formula:
C₂₁H₂₇N₇Na₂O₁₄P₂ MW: 709.41 g/mol |
MDL Number:
MFCD00036200 CAS Number: 606-68-8 EINECS: 210-123-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...