β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Nicotinamide adenine dinucleotide, oxidizedNadideNAD+

SIALN8285-15VLEA 348 GBP
SIALN8285-15VL
β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form), Sigma-Aldrich®
β-Nicotinamide adenine dinucleotide (NAD, oxidized form)
Formula: C₂₁H₂₇N₇O₁₄P₂
MW: 663.43 g/mol
Melting Pt: 140 to 142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036253
CAS Number: 53-84-9
EINECS: 200-184-4
Merck Index: 12,06429

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR