Flash chromatography columns, PuriFlash®, with MM1 silica
Supplier: Interchim
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PuriFlash®
INTEPF50MM1/55GEA 154 GBP
INTEPF50MM1/55G INTEPF50MM1/35G INTEPF50MM1/6G INTEPF50MM1/20G INTEPF50MM1/2500G INTEPF50MM1/450G INTEPF50MM1/175G INTEPF50MM1/120G INTEPF50MM1/300G INTEPF50MM1/1250G
Flash chromatography columns, PuriFlash®, with MM1 silica
Chromatography Columns
These flash chromatography columns with ion exchange bonded MM1 silica are used for unique selectivity of basic, charged and non-polar compounds via ion exchange and hydrophobic chains (SCX/C8).
Compounds with basic functionality are retained by ion exchange. A washing step with an appropriate pH, elutes ionizable compounds. Retained compounds from hydrophobic bonding can be eluted with an organic solvent.