Flash chromatography columns, PuriFlash®, C18 HP silica
Supplier: Interchim
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PuriFlash®
INTEPF30C18HP/6GEA 96 GBP
INTEPF30C18HP/6G INTEPF30C18HP/35G INTEPF15C18HP/6G INTEPF50C18HP/35G INTEPF50C18HP/55G INTEPF50C18HP/2500 INTEPF50C18HP/1250 INTEPF30C18HP/300G INTEPF50C18HP/175G INTEPF50C18HP/450G INTEPF30C18HP/120G INTEPF15C18HP/175G INTEPF15C18HP/450G INTEPF15C18HP/20G INTEPF30C18HP/20G INTEPF30C18HP/2500 INTEPF50C18HP/20G INTEPF30C18HP/1250 INTEPF15C18HP/2500 INTEPF50C18HP/120G INTEPF30C18HP/450G INTEPF15C18HP/1250 INTEPF50C18HP/300G INTEPF30C18HP/175G INTEPF15C18HP/55G INTEPF15C18HP/120G INTEPF15C18HP/35G INTEPF30C18HP/55G INTEPF50C18HP/6G INTEPF15C18HP/300G
Flash chromatography columns, PuriFlash®, C18 HP silica
Chromatography Columns
These flash chromatography columns, C18 HP silica are suitable for the purification of non polar and ionic compounds.
  • Base material is high performance spherical silica
  • pH stability ranges from 1 to 7,5