Flash chromatography columns, PuriFlash®, Cellufine™ Etclean L
Supplier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CF90EL/3100ML CF90EL/7ML CF90EL/68ML CF90EL/575ML CF90EL/375ML CF90EL/27ML CF90EL/215ML CF90EL/1540ML CF90EL/145ML CF90EL/45ML
INTECF90EL/3100MLEA 0 GBP
INTECF90EL/3100ML INTECF90EL/7ML INTECF90EL/68ML INTECF90EL/575ML INTECF90EL/375ML INTECF90EL/27ML INTECF90EL/215ML INTECF90EL/1540ML INTECF90EL/145ML INTECF90EL/45ML
Flash chromatography columns, PuriFlash®, Cellufine™ Etclean L
Chromatography Columns
These flash chromatography columns have immobilised poly (e-lysine) Cellufine™ beads for affinity, endotoxin removal up to 2000 kD.
  • pH stability from 2 to 13
  • Removes 480 µg endotoxine/ml