Flash chromatography columns, PuriFlash®, Cellufine™ Etclean S
Supplier: Interchim
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PuriFlash®
INTECF75ES/3100MLEA 54380 GBP
INTECF75ES/3100ML INTECF75ES/375ML INTECF75ES/7ML INTECF75ES/575ML INTECF75ES/27ML INTECF75ES/1540ML INTECF75ES/68ML INTECF75ES/215ML INTECF75ES/45ML INTECF75ES/145ML
Flash chromatography columns, PuriFlash®, Cellufine™ Etclean S
Chromatography Columns
These flash chromatography columns have immobilised poly (e-lysine) Cellufine™ beads for affinity, endotoxin removal up to 2 kD.
  • pH stability from 2 to 13
  • Removes 185 µg endotoxine/ml