Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt

ARC-1546 ARC-1546-1 ARC-1546-2
BTRNARC-1546EA 0 GBP
BTRNARC-1546 BTRNARC-1546-1 BTRNARC-1546-2
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)
Formula: C₉H₁₂N₂Na₃O₁₅P₃
MW: 550.09 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00044310
CAS Number: 19817-92-6
EINECS: 243-347-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR