5-Iodo-2'-deoxyuridine (Idoxuridine)

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dioneIdoxuridine(+)-5-Iodo-2'-deoxyuridine

MOLEM19199640EA 0 GBP
MOLEM19199640
5-Iodo-2'-deoxyuridine (Idoxuridine)
5-Iodo-2'-deoxyuridine (Idoxuridine)
Formula: C₉H₁₁IN₂O₅
MW: 354.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134656
CAS Number: 54-42-2
EINECS: 200-207-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR