Kanamycin sulphate 50 mg/ml in solution, Ultra Pure Grade

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Kanamycin Monosulphate(2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphateKanamycin A

E713-20MLEA 123 GBP
E713-20ML
Kanamycin sulphate 50 mg/ml in solution, Ultra Pure Grade
Kanamycin sulphate

Kanamycin is an aminoglycoside antibiotic that inhibits protein synthesis in gram-negative and gram-positive bacteria and in mycoplasma. Recommended working concentration: 30 µg/ml.

Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
MDL Number: MFCD00070253
CAS Number: 25389-94-0

Order Now

Specification Test Results

Clarity PASS / Fail
Specific Gravity (20 °C) 1,0186 - 1,0427

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR