2-(Tributylstannyl)pyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-(Tri-n-butylstannyl)pyridine

M-Clarity™ quality level = MQ100

SIAL678333-1GEA 79 GBP
SIAL678333-1G SIAL678333-5G
2-(Tributylstannyl)pyridine, Sigma-Aldrich®
2-(Tributylstannyl)pyridine
Formula: C₁₇H₃₁NSn
MW: 368.15 g/mol
MDL Number: MFCD00052052
CAS Number: 17997-47-6
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR