2-Methoxy-6-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS50205G1 AS50205G5
ACTIAS50205G1EA 132 GBP
ACTIAS50205G1 ACTIAS50205G5
2-Methoxy-6-(tributylstannyl)pyridine
2-Methoxy-6-(tributylstannyl)pyridine
Formula: C₁₈H₃₃NOSn
MDL Number: MFCD09025803
CAS Number: 164014-94-2
UN: 2788
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR