2-Fluoro-4-(tributylstannyl)pyridine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
719315-1G
719315-5G
SIAL719315-1GEA
172
GBP
SIAL719315-1G
SIAL719315-5G
2-Fluoro-4-(tributylstannyl)pyridine, Sigma-Aldrich®
2-Fluoro-4-(tributylstannyl)pyridine
Formula:
C₁₇H₃₀FNSn |
CAS Number:
457061-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...