(±)-Indoline-2-carboxylic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Indoline-2-carboxylic acid

M-Clarity™ quality level = MQ100

302244-25G 302244-5G
SIAL302244-25GEA 132 GBP
SIAL302244-25G SIAL302244-5G
(±)-Indoline-2-carboxylic acid, Sigma-Aldrich®
(±)-Indoline-2-carboxylic acid
Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00010635
CAS Number: 78348-24-0
EINECS: 278-899-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR