2-Amino-3-thiophenecarbonitrile, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Aminothiophene-3-carbonitrile

681067-1G 681067-5G
SIAL681067-1GEA 52.6 GBP
SIAL681067-1G SIAL681067-5G
2-Amino-3-thiophenecarbonitrile, Sigma-Aldrich®
2-Amino-3-thiophenecarbonitrile
Formula: C₅H₄N₂S
MW: 124.16 g/mol
MDL Number: MFCD00706298
CAS Number: 4651-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR