2-Amino-3-thiophenecarbonitrile

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Aminothiophene-3-carbonitrile

4N-807.500 4N-807.1 4N-807.5 4N-807.10
BIOT4N-807.500EA 0 GBP
BIOT4N-807.500 BIOT4N-807.1 BIOT4N-807.5 BIOT4N-807.10
2-Amino-3-thiophenecarbonitrile
2-Amino-3-thiophenecarbonitrile
Formula: C₅H₄N₂S
MW: 124.16 g/mol
MDL Number: MFCD00706298
CAS Number: 4651-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR