1,3-Dichloro-6-methoxyisoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6018-5G HC-6018-1G
COBBHC-6018-5GEA 0 GBP
COBBHC-6018-5G COBBHC-6018-1G
1,3-Dichloro-6-methoxyisoquinoline
1,3-Dichloro-6-methoxyisoquinoline
Formula: C₁₀H₇Cl₂NO
MW: 228.747 g/mol
MDL Number: MFCD09258765
CAS Number: 24623-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR