Cupferron

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ammonium 2-oxo-1-phenylhydrazinolate

MOLEM31862784EA 0 GBP
MOLEM31862784
Cupferron
Cupferron
Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
MDL Number: MFCD00078422
CAS Number: 135-20-6
EINECS: 205-183-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR