Cupferron ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Ammonium 2-oxo-1-phenylhydrazinolate
A16551.14EA
19.1
GBP
A16551.14
Cupferron ≥97%
Cupferron
Formula:
C₆H₉N₃O₂ MW: 155.16 g/mol Melting Pt: 154 °C (dec.) |
MDL Number:
MFCD00078422 CAS Number: 135-20-6 EINECS: 205-183-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...