Choline hydrogen tartrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: CBT2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoateCholine bitartrate2-(Hydroxyethyl)trimethylammonium bitartrateCholine acid tartrate

M-Clarity™ quality level = MQ200

SIALC1629-500GEA 223 GBP
SIALC1629-500G PFIZC-1629
Choline hydrogen tartrate, Sigma-Aldrich®
Choline hydrogen tartrate
Formula: C₉H₁₉NO₇
MW: 253.25 g/mol
MDL Number: MFCD00036332
CAS Number: 87-67-2
EINECS: 201-763-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR