Pebulate(N-butyl-N-ethyl-S-propylthiocarbamate)

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

EHERC15904000EA 203 GBP
EHERC15904000
Pebulate(N-butyl-N-ethyl-S-propylthiocarbamate)
Pebulate (N-Butyl-N-ethyl-S-propylthiocarbamate)
Formula: C₁₀H₂₁NOS
CAS Number: 1114-71-2
EINECS: 214-215-4
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR