(S)-(-)-2-Pyrrolidinone-5-carboxylic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (S)-(-)-2-Pyrrolidone-5-carboxylic acidL-Pyroglutamic Acid(S)-5-Oxopyrrolidine-2-carboxylic acidH-Pyr-OH

M-Clarity™ quality level = MQ200

SIALP5960-25GEA 114 GBP
SIALP5960-25G
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid, Sigma-Aldrich®
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid
Formula: C₅H₇NO₃
MW: 129.1 g/mol
MDL Number: MFCD00005272
CAS Number: 98-79-3
EINECS: 202-700-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR