5-Fluoroorotic acid

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-FOA5-Fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid5-Fluorouracil-4-carboxylic acid2,6-Dihydroxy-5-fluoropyrimidine-4-carboxylic acid

FLUOM02721-5GEA 0 GBP
FLUOM02721-5G
5-Fluoroorotic acid
5-Fluoroorotic acid
Formula: C₅H₃FN₂O₄
MW: 174.09 g/mol
MDL Number: MFCD00042526
CAS Number: 703-95-7
EINECS: 211-876-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR