5-Fluoroorotic acid
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-FOA
, 5-Fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
, 5-Fluorouracil-4-carboxylic acid
, 2,6-Dihydroxy-5-fluoropyrimidine-4-carboxylic acid
ROTH4015.3EA
130.5
GBP
ROTH4015.3
5-Fluoroorotic acid
5-Fluoroorotic acid
Formula:
C₅H₃FN₂O₄ MW: 174.09 g/mol |
MDL Number:
MFCD00042526 CAS Number: 703-95-7 EINECS: 211-876-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...