Nickel carbonate, basic hydrate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
544183-1KG-A
544183-250G-A
SIAL544183-1KG-AEA
230
GBP
SIAL544183-1KG-A
SIAL544183-250G-A
Nickel carbonate, basic hydrate, Sigma-Aldrich®
Nickel carbonate, basic hydrate
Formula:
NiCO₃·2Ni(OH)₂·xH₂O MW: 304.12 g/mol (anhy) |
MDL Number:
MFCD00150262 CAS Number: 958638-02-3 EINECS: 235-715-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...