4-(2-Bromoethoxy)benzaldehyde

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

3G-938.5 3G-938.500 3G-938.1 3G-938.10
BIOT3G-938.5EA 0 GBP
BIOT3G-938.5 BIOT3G-938.500 BIOT3G-938.1 BIOT3G-938.10
4-(2-Bromoethoxy)benzaldehyde
4-(2-Bromoethoxy)benzaldehyde
Formula: C₉H₉BrO₂
MW: 229.0706 g/mol
MDL Number: MFCD00021074
CAS Number: 52191-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR