3-Chloro-2-fluorophenylacetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005669-1G 005669-5G
FLUO005669-1GEA 0 GBP
FLUO005669-1G FLUO005669-5G
3-Chloro-2-fluorophenylacetonitrile
3-Chloro-2-fluorophenylacetonitrile
Formula: C₈H₅ClFN
MW: 169.5800 g/mol
MDL Number: MFCD01631552
CAS Number: 261762-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR