Fluorene-1-carboxylic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

SIALF1204-1GEA 97.3 GBP
SIALF1204-1G
Fluorene-1-carboxylic acid, Sigma-Aldrich®
Fluorene-1-carboxylic acid
Formula: C₁₄H₁₀O₂
MW: 210.23 g/mol
MDL Number: MFCD00001130
CAS Number: 6276-03-5
EINECS: 228-472-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR