Fluorene-1-carboxylic acid
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MOLE28108738-1GEA
0
GBP
MOLE28108738-1G
Fluorene-1-carboxylic acid
Fluorene-1-carboxylic acid
Formula:
C₁₄H₁₀O₂ MW: 210.23 g/mol |
MDL Number:
MFCD00001130 CAS Number: 6276-03-5 EINECS: 228-472-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...