2,5-Dibromo-3-butylthiophene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

525499-5G 525499-1G
SIAL525499-5GEA 720 GBP
SIAL525499-5G SIAL525499-1G
2,5-Dibromo-3-butylthiophene, Sigma-Aldrich®
2,5-Dibromo-3-butylthiophene
Formula: C₈H₁₀Br₂S
MW: 298.04 g/mol
MDL Number: MFCD00672012
CAS Number: 116971-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR