2-Fluoro-4-methoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-4-methoxybenzeneboronic acid

BB-2573-25G BB-2573-5G BB-2573-1G
COBBBB-2573-25GEA 0 GBP
COBBBB-2573-25G COBBBB-2573-5G COBBBB-2573-1G
2-Fluoro-4-methoxyphenylboronic acid
2-Fluoro-4-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD04039323
CAS Number: 162101-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR