1-Methyl-1-phenylhydrazine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

170267-25G 170267-5G
SIAL170267-25GEA 489 GBP
SIAL170267-25G SIAL170267-5G
1-Methyl-1-phenylhydrazine, Sigma-Aldrich®
1-Methyl-1-phenylhydrazine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00007627
CAS Number: 618-40-6
EINECS: 210-547-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR