N-Methyl-2,2,2-trifluoroacetamide

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-Methyltrifluoroacetamide, 2,2,2-Trifluoro-N-methylacetamide

009293-25G 009293-100G
FLUO009293-25GEA 0 GBP
FLUO009293-25G FLUO009293-100G
N-Methyl-2,2,2-trifluoroacetamide
N-Methyl-2,2,2-trifluoroacetamide
Formula: C₃H₄F₃NO
MW: 127.07 g/mol
MDL Number: MFCD00009670
CAS Number: 815-06-5
EINECS: 212-417-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR