2-Bromo-N-(2,6-dimethylphenyl)acetamide

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

1G-022.500 1G-022.1 1G-022.5 1G-022.10
BIOT1G-022.500EA 0 GBP
BIOT1G-022.500 BIOT1G-022.1 BIOT1G-022.5 BIOT1G-022.10
2-Bromo-N-(2,6-dimethylphenyl)acetamide
2-Bromo-N-(2,6-dimethylphenyl)acetamide
Formula: C₁₀H₁₂BrNO
MW: 242.12 g/mol
MDL Number: MFCD00215011
CAS Number: 32433-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR