Cacodylic acid (dimethylarsinic acid)
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Dimethylarsinic acid (Cacodylic acid)
MOLEM12133684EA
0
GBP
MOLEM12133684
Cacodylic acid (dimethylarsinic acid)
Cacodylic acid (Dimethylarsinic acid)
Formula:
(CH₃)₂As(O)OH MW: 138 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002095 CAS Number: 75-60-5 EINECS: 200-883-4 UN: 1572 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...