Ketoprofen, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(3-Benzoylphenyl)propanoic acid2-(3-Benzoylphenyl)propionic acid

M-Clarity™ quality level = MQ100

34016-100MG-REA 57.9 GBP
34016-100MG-R
Ketoprofen, Supelco®
Ketoprofen
Formula: C₁₆H₁₄O₃
MW: 254.29 g/mol
MDL Number: MFCD00055790
CAS Number: 22071-15-4
EINECS: 244-759-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR