Ketoprofen
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(3-Benzoylphenyl)propanoic acid
, 2-(3-Benzoylphenyl)propionic acid
MOLE41476439-5GEA
0
GBP
MOLE41476439-5G
Ketoprofen
Ketoprofen
Formula:
C₁₆H₁₄O₃ MW: 254.29 g/mol |
MDL Number:
MFCD00055790 CAS Number: 22071-15-4 EINECS: 244-759-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...