4-Chloro-4'-hydroxybutyrophenone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

C44907-25G C44907-5G
SIALC44907-25GEA 296 GBP
SIALC44907-25G SIALC44907-5G
4-Chloro-4'-hydroxybutyrophenone, Sigma-Aldrich®
4-Chloro-4'-hydroxybutyrophenone
Formula: C₁₀H₁₁ClO₂
MDL Number: MFCD00002362
CAS Number: 7150-55-2
EINECS: 230-485-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR