Dibutylamine 260 g/l ∼2 mol/l in toluene isocyanate reaction solution
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Di-n-butylamine, N-butyl-1-propanamine
16571.4700
16571.2700
16571.3700
KRAF16571.4700EA
0
GBP
KRAF16571.4700
KRAF16571.2700
KRAF16571.3700
Dibutylamine 260 g/l ∼2 mol/l in toluene isocyanate reaction solution
Dibutylamine
Formula:
C₈H₁₉N MW: 129.25 g/mol |
MDL Number:
MFCD00009429 CAS Number: 111-92-2 EINECS: 203-921-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...