Silicone oil
Supplier: Hach
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
126936
HACH126936EA
21.2
GBP
HACH126936
Silicone oil
Silicone oil
Multiple uses, including preparation of turbidity sample cells.
Density:
0.9…1.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00132673 CAS Number: 68083-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...