1,2,3,4-Butanetetracarboxylic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ100
257303-100G
257303-500G
SIAL257303-100GEA
143
GBP
SIAL257303-100G
SIAL257303-500G
1,2,3,4-Butanetetracarboxylic acid, Sigma-Aldrich®
1,2,3,4-Butanetetracarboxylic acid
Formula:
C₈H₁₀O₈ MW: 234.16 g/mol Boiling Pt: 460 °C (1013 hPa) Melting Pt: 189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002722 CAS Number: 1703-58-8 EINECS: 216-938-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...